Ethyl-3-amino-1-benzothiophene-2-carboxylate
Supplier: Matrix Scientific
080313-500MG
080313-1G
080313-5G
102669-034EA
161.54
USD
102669-034
102669-036
102669-038
Ethyl-3-amino-1-benzothiophene-2-carboxylate
Ethyl-3-amino-1-benzothiophene-2-carboxylate
Hazard: Irritant
Formula:
C₁₁H₁₁NO₂S MW: 221.28 g/mol |
MDL Number:
MFCD00661903 CAS Number: 34761-09-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...