[2-(3-Nitro-1H-pyrazol-1-yl)ethyl]amine hydrochloride

Supplier: Matrix Scientific

080308-500MG 080308-1G 080308-5G
102669-004EA 646.15 USD
102669-004 102669-006 102669-008
[2-(3-Nitro-1H-pyrazol-1-yl)ethyl]amine hydrochloride
[2-(3-Nitro-1H-pyrazol-1-yl)ethyl]amine hydrochloride

Hazard: Irritant

Formula: C₅H₉ClN₄O₂
MDL Number: MFCD15142894
CAS Number: 1197235-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR