N-(2-Aminoethyl)-5-nitro-1H-pyrazole-3-carboxamide

Supplier: Matrix Scientific

080292-500MG 080292-5G
102668-908EA 646.15 USD
102668-908 102668-912
N-(2-Aminoethyl)-5-nitro-1H-pyrazole-3-carboxamide
N-(2-Aminoethyl)-5-nitro-1H-pyrazole-3-carboxamide

Hazard: Irritant

Formula: C₆H₉N₅O₃
MDL Number: MFCD05862158
CAS Number: 1159989-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR