3-(Methylthio)-5-(2-thienyl)-4H-1,2,4-triazol-4-amine

Supplier: Matrix Scientific

080290-500MG 080290-5G
102668-896EA 387.69 USD
102668-896 102668-900
3-(Methylthio)-5-(2-thienyl)-4H-1,2,4-triazol-4-amine
3-(Methylthio)-5-(2-thienyl)-4H-1,2,4-triazol-4-amine

Hazard: Irritant

Formula: C₇H₈N₄S₂
MDL Number: MFCD01465907
CAS Number: 118158-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR