1-(2,2,2-Trifluoroethyl)-1H-pyrazol-4-amine hydrochloride
Supplier: Matrix Scientific
080282-500MG
080282-5G
102668-848EA
553.85
USD
102668-848
102668-852
1-(2,2,2-Trifluoroethyl)-1H-pyrazol-4-amine hydrochloride
1-(2,2,2-Trifluoroethyl)-1H-pyrazol-4-amine hydrochloride
Hazard: Irritant
Formula:
C₅H₇ClF₃N₃ MW: 201.58 g/mol |
MDL Number:
MFCD04967266 CAS Number: 1185177-94-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...