1,3-Dimethyl-6-(2-thienyl)-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

Supplier: Matrix Scientific

080270-500MG 080270-5G
102668-776EA 258.46 USD
102668-776 102668-780
1,3-Dimethyl-6-(2-thienyl)-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid
1,3-Dimethyl-6-(2-thienyl)-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₁N₃O₂S
MDL Number: MFCD08558202
CAS Number: 893645-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR