7-(3,4-Dimethoxyphenyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

Supplier: Matrix Scientific

080266-500MG 080266-5G
102668-752EA 387.69 USD
102668-752 102668-756
7-(3,4-Dimethoxyphenyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid
7-(3,4-Dimethoxyphenyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₃N₃O₄
MDL Number: MFCD06739362
CAS Number: 886503-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR