(5-Methyl-4-nitro-1H-pyrazol-1-yl)acetic acid

Supplier: Matrix Scientific

080254-500MG 080254-5G
102668-680EA 646.15 USD
102668-680 102668-684
(5-Methyl-4-nitro-1H-pyrazol-1-yl)acetic acid
(5-Methyl-4-nitro-1H-pyrazol-1-yl)acetic acid

Hazard: Irritant

Formula: C₆H₇N₃O₄
MDL Number: MFCD04967124
CAS Number: 1001611-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR