4-Nitro-1H-pyrazole-5-carbonitrile
Supplier: Matrix Scientific
Synonyms:
4-Nitro-1H-pyrazole-3-carbonitrile
080249-500MG
080249-5G
102668-652EA
646.15
USD
102668-652
102668-656
4-Nitro-1H-pyrazole-5-carbonitrile
4-Nitro-1H-pyrazole-5-carbonitrile
Hazard: Irritant
Formula:
C₄H₂N₄O₂ MW: 138.09 g/mol |
MDL Number:
MFCD15143246 CAS Number: 61241-07-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...