5-Bromo-1,3-dimethyl-1H-1,2,4-triazole

Supplier: Matrix Scientific

080235-500MG 080235-5G
102668-578EA 646.15 USD
102668-578 102668-582
5-Bromo-1,3-dimethyl-1H-1,2,4-triazole
5-Bromo-1,3-dimethyl-1H-1,2,4-triazole

Hazard: Irritant

Formula: C₄H₆BrN₃
MDL Number: MFCD22066399
CAS Number: 56616-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR