[1-(Methoxymethyl)cyclobutyl]amine hydrochloride

Supplier: Matrix Scientific

080228-500MG 080228-5G
102668-536EA 646.15 USD
102668-536 102668-540
[1-(Methoxymethyl)cyclobutyl]amine hydrochloride
[1-(Methoxymethyl)cyclobutyl]amine hydrochloride

Hazard: Irritant

Formula: C₆H₁₄ClNO
MDL Number: MFCD15474849
CAS Number: 1220039-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR