1-(1,1-Dimethylpropyl)-5-oxopyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

080210-500MG 080210-5G
102668-448EA 516.92 USD
102668-448 102668-452
1-(1,1-Dimethylpropyl)-5-oxopyrrolidine-3-carboxylic acid
1-(1,1-Dimethylpropyl)-5-oxopyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₇NO₃
MDL Number: MFCD07364765
CAS Number: 944683-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR