1-(2-Aminoethyl)piperidine

Supplier: Matrix Scientific

Synonyms: N-(2-Aminoethyl)piperidine

079892-5G 079892-25G
102833-958EA 55.38 USD
102833-958 102833-960
1-(2-Aminoethyl)piperidine
1-(2-Aminoethyl)piperidine

Hazard: Irritant

Formula: C₇H₁₆N₂
MDL Number: MFCD00006516
CAS Number: 27578-60-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR