2-Chloro-4-methyl-6-nitroquinoline

Supplier: Matrix Scientific

079496-500MG 079496-5G
102668-408EA 335.38 USD
102668-408 102668-412
2-Chloro-4-methyl-6-nitroquinoline
2-Chloro-4-methyl-6-nitroquinoline

Hazard: Irritant

Formula: C₁₀H₇ClN₂O₂
MDL Number: MFCD00169016
CAS Number: 54965-59-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR