N-Ethyl-p-toluidine

Supplier: Matrix Scientific

079249-1G 079249-5G
102833-950EA 267.69 USD
102833-950 102833-952
N-Ethyl-p-toluidine
N-Ethyl-p-toluidine

Hazard: Toxic/Stored under Argon

Formula: C₉H₁₃N
Boiling Pt: 222 °C
Density: 0.94
MDL Number: MFCD00035793
CAS Number: 622-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR