8-hydroxyquinoline-2-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 8-Hydroxyquinoline-2-carboxylic acid

079133-500MG 079133-5G
102668-396EA 387.69 USD
102668-396 102668-400
8-hydroxyquinoline-2-carboxylic acid
8-hydroxyquinoline-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
MDL Number: MFCD00168972
CAS Number: 1571-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR