Ethyl cyano(hydroxyimino)acetate

Supplier: Matrix Scientific

Synonyms: Ethyl 2-cyano-2-(hydroxyimino)acetate, Ethyl Cyanoglyoxalate Oxime, Cyano(hydroxyimino)acetic Acid Ethyl Ester, Oxyma

078476-100G 078476-500G
102833-946EA 67.69 USD
102833-946 102833-948
Ethyl cyano(hydroxyimino)acetate
Ethyl cyano(hydroxyimino)acetate

Hazard: Irritant/Refrigerate

Formula: C₅H₆N₂O₃
Melting Pt: 127…132 °C
MDL Number: MFCD00002112
CAS Number: 3849-21-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR