Levoglucosenone
Supplier: Matrix Scientific
Synonyms:
1,6-anhydro-3,4-dideoxyhex-3-enopyran-2-ulose, 6,8-Dioxabicyclo[3.2.1]oct-2-en-4-one, (1S,5R)-6,8-Dioxabicyclo[3.2.1]oct-2-en-4-one
078186-500MG
078186-5G
102668-350EA
958.46
USD
102668-350
102668-354
Levoglucosenone
Levoglucosenone
CAS: 37112-31-5
MDL Number: MFCD19237758
Molecular Formula: C6H6O3
Molecular Weight: 126,11
Hazard: Irritant
Formula:
C₆H₆O₃ MW: 126.11 g/mol |
MDL Number:
MFCD00191664 CAS Number: 37112-31-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...