N,N-Dimethylvaline hydrochloride

Supplier: Matrix Scientific

Synonyms: N,N-Dimethyvaline

077879-500MG 077879-5G
102668-160EA 387.69 USD
102668-160 102668-164
N,N-Dimethylvaline hydrochloride
N,N-Dimethylvaline hydrochloride

Hazard: Irritant

Formula: C₇H₁₆ClNO₂
MW: 181.66 g/mol
MDL Number: MFCD22123248
CAS Number: 2812-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR