N-(3-Methoxybenzoyl)-β-alanine

Supplier: Matrix Scientific

077858-500MG 077858-5G
102668-062EA 516.92 USD
102668-062 102668-066
N-(3-Methoxybenzoyl)-β-alanine
N-(3-Methoxybenzoyl)-beta-alanine

Hazard: Irritant

Formula: C₁₁H₁₃NO₄
MW: 223.23 g/mol
MDL Number: MFCD08444965
CAS Number: 914773-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR