Pyrimidine-4,5,6-triamine

Supplier: Matrix Scientific

077838-500MG 077838-5G
102667-994EA 161.54 USD
102667-994 102667-998
Pyrimidine-4,5,6-triamine
Pyrimidine-4,5,6-triamine

Hazard: Irritant

Formula: C₄H₇N₅
MDL Number: MFCD00023271
CAS Number: 118-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR