3-(Chloromethyl)-5-cyclopropyl-1,2,4-oxadiazole

Supplier: Matrix Scientific

077816-500MG 077816-5G
102667-896EA 516.92 USD
102667-896 102667-900
3-(Chloromethyl)-5-cyclopropyl-1,2,4-oxadiazole
3-(Chloromethyl)-5-cyclopropyl-1,2,4-oxadiazole

Hazard: Irritant

Formula: C₆H₇ClN₂O
MDL Number: MFCD09864335
CAS Number: 959237-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR