5-Phenyl-1,2,4-oxadiazol-3-amine
Supplier: Matrix Scientific
077814-500MG
077814-5G
102667-884EA
369.23
USD
102667-884
102667-888
5-Phenyl-1,2,4-oxadiazol-3-amine
5-Phenyl-1,2,4-oxadiazol-3-amine
Hazard: Irritant
Formula:
C₈H₇N₃O |
MDL Number:
MFCD00197235 CAS Number: 7788-14-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...