1-(Dimethylamino)cyclopentanecarbonitrile

Supplier: Matrix Scientific

077801-500MG 077801-5G
102667-822EA 516.92 USD
102667-822 102667-826
1-(Dimethylamino)cyclopentanecarbonitrile
1-(Dimethylamino)cyclopentanecarbonitrile

Hazard: Irritant

Formula: C₈H₁₄N₂
MDL Number: MFCD09880530
CAS Number: 22912-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR