3-(Chloromethyl)-5-cyclobutyl-1,2,4-oxadiazole

Supplier: Matrix Scientific

077797-500MG 077797-5G
102667-798EA 646.15 USD
102667-798 102667-802
3-(Chloromethyl)-5-cyclobutyl-1,2,4-oxadiazole
3-(Chloromethyl)-5-cyclobutyl-1,2,4-oxadiazole

Hazard: Irritant

Formula: C₇H₉ClN₂O
MDL Number: MFCD00728735
CAS Number: 259683-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR