1,5-Dimethyl-3-nitro-1H-pyrazole
Supplier: Matrix Scientific
077763-500MG
077763-5G
102667-616EA
646.15
USD
102667-616
102667-620
1,5-Dimethyl-3-nitro-1H-pyrazole
1,5-Dimethyl-3-nitro-1H-pyrazole
Hazard: Irritant
Formula:
C₅H₇N₃O₂ |
MDL Number:
MFCD04971109 CAS Number: 59376-16-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...