(2-Phenyl-1,3-thiazol-5-yl)methanol

Supplier: Matrix Scientific

Synonyms: (2-Phenylthiazol-5-yl)methanol

077663-500MG 077663-1G 077663-5G
102667-358EA 369.23 USD
102667-358 102667-360 102667-362
(2-Phenyl-1,3-thiazol-5-yl)methanol
(2-Phenyl-1,3-thiazol-5-yl)methanol

Hazard: Irritant

Formula: C₁₀H₉NOS
MW: 191.25 g/mol
MDL Number: MFCD09694149
CAS Number: 859485-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR