4-Amino-1-phenylpyrrolidin-2-one hydrochloride

Supplier: Matrix Scientific

077653-500MG 077653-1G 077653-5G
102667-330EA 492.31 USD
102667-330 102667-332 102667-334
4-Amino-1-phenylpyrrolidin-2-one hydrochloride
4-Amino-1-phenylpyrrolidin-2-one hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O
MW: 212.68 g/mol
MDL Number: MFCD01687393
CAS Number: 3026-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR