2-Ethoxy-2-methylpropanoic acid

Supplier: Matrix Scientific

077651-500MG 077651-1G 077651-5G
102667-320EA 360 USD
102667-320 102667-322 102667-324
2-Ethoxy-2-methylpropanoic acid
2-Ethoxy-2-methylpropanoic acid

Hazard: Irritant

Formula: C₆H₁₂O₃
MDL Number: MFCD14636712
CAS Number: 15001-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR