4-(Chloromethyl)-2-(3,4-dimethoxyphenyl)-5-methyl-1,3-oxazole

Supplier: Matrix Scientific

077650-500MG 077650-1G 077650-5G
102667-314EA 516.92 USD
102667-314 102667-316 102667-318
4-(Chloromethyl)-2-(3,4-dimethoxyphenyl)-5-methyl-1,3-oxazole
4-(Chloromethyl)-2-(3,4-dimethoxyphenyl)-5-methyl-1,3-oxazole

Hazard: Irritant

Formula: C₁₃H₁₄ClNO₃
MDL Number: MFCD08457266
CAS Number: 907200-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR