(3,4-Difluorophenoxy)acetic acid

Supplier: Matrix Scientific

077649-500MG 077649-1G
102667-310EA 133.85 USD
102667-310 102667-312
(3,4-Difluorophenoxy)acetic acid
(3,4-Difluorophenoxy)acetic acid

Hazard: Irritant

Formula: C₈H₆F₂O₃
MDL Number: MFCD06823827
CAS Number: 370-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR