5-Formylsalicylic acid

Supplier: Matrix Scientific

077638-500MG 077638-1G 077638-5G
102667-248EA 15.38 USD
102667-248 102667-250 102667-252
5-Formylsalicylic acid
5-Formylsalicylic acid

Hazard: Irritant

Formula: C₈H₆O₄
MDL Number: MFCD00006945
CAS Number: 616-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR