Ethyl-1-ethyl-1H-pyrazole-3-carboxylate

Supplier: Matrix Scientific

077637-500MG 077637-1G 077637-5G
102667-242EA 387.69 USD
102667-242 102667-244 102667-246
Ethyl-1-ethyl-1H-pyrazole-3-carboxylate
Ethyl-1-ethyl-1H-pyrazole-3-carboxylate

Hazard: Irritant

Formula: C₈H₁₂N₂O₂
MDL Number: MFCD08700856
CAS Number: 1007503-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR