2-Hydroxy-6-methyl-1,2,3,4-tetrahydroquinoline-4-carboxylic acid

Supplier: Matrix Scientific

077616-500MG 077616-1G 077616-5G
102667-128EA 646.15 USD
102667-128 102667-130 102667-132
2-Hydroxy-6-methyl-1,2,3,4-tetrahydroquinoline-4-carboxylic acid
2-Hydroxy-6-methyl-1,2,3,4-tetrahydroquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₃
MDL Number: MFCD20441393
CAS Number: 1338495-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR