5-Bromo-1-ethyl-1H-1,2,4-triazole

Supplier: Matrix Scientific

077608-500MG 077608-1G 077608-5G
102667-080EA 516.92 USD
102667-080 102667-082 102667-084
5-Bromo-1-ethyl-1H-1,2,4-triazole
5-Bromo-1-ethyl-1H-1,2,4-triazole

Hazard: Irritant

Formula: C₄H₆BrN₃
MDL Number: MFCD16987772
CAS Number: 64907-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR