Dibenzo[b,d]furan-2-ol

Supplier: Matrix Scientific

077607-500MG 077607-1G
102667-076EA 387.69 USD
102667-076 102667-078
Dibenzo[b,d]furan-2-ol
Dibenzo[b,d]furan-2-ol

Hazard: Irritant

Formula: C₁₂H₈O₂
MDL Number: MFCD00010325
CAS Number: 86-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR