2-Chloro-5-fluoro-N-methylpyrimidin-4-amine

Supplier: Matrix Scientific

077601-500MG 077601-1G 077601-5G
102667-052EA 646.15 USD
102667-052 102667-054 102667-056
2-Chloro-5-fluoro-N-methylpyrimidin-4-amine
2-Chloro-5-fluoro-N-methylpyrimidin-4-amine

Hazard: Irritant

Formula: C₅H₅ClFN₃
MDL Number: MFCD01346699
CAS Number: 67316-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR