2-Chloro-5-fluoro-N-phenylpyrimidin-4-amine

Supplier: Matrix Scientific

077600-500MG 077600-1G 077600-5G
102667-046EA 387.69 USD
102667-046 102667-048 102667-050
2-Chloro-5-fluoro-N-phenylpyrimidin-4-amine
2-Chloro-5-fluoro-N-phenylpyrimidin-4-amine

Hazard: Irritant

Formula: C₁₀H₇ClFN₃
MDL Number: MFCD20441387
CAS Number: 280581-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR