3-(1H-1,2,4-Triazol-3-yl)pyridine

Supplier: Matrix Scientific

077584-500MG 077584-1G
102666-994EA 516.92 USD
102666-994 102666-996
3-(1H-1,2,4-Triazol-3-yl)pyridine
3-(1H-1,2,4-Triazol-3-yl)pyridine

Hazard: Irritant

Formula: C₇H₆N₄
MDL Number: MFCD18260902
CAS Number: 23195-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR