4-(1H-1,2,4-Triazol-3-yl)pyridine

Supplier: Matrix Scientific

077583-500MG 077583-1G
102666-990EA 516.92 USD
102666-990 102666-992
4-(1H-1,2,4-Triazol-3-yl)pyridine
4-(1H-1,2,4-Triazol-3-yl)pyridine

Hazard: Irritant

Formula: C₇H₆N₄
MDL Number: MFCD18260903
CAS Number: 14803-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR