2-Chloro-5-hydrazinylpyridine

Supplier: Matrix Scientific

Synonyms: 2-Chloro-5-hydrazinylpyridine

077582-500MG 077582-1G
102666-986EA 646.15 USD
102666-986 102666-988
2-Chloro-5-hydrazinylpyridine
2-Chloro-5-hydrazinylpyridine

Hazard: Irritant

Formula: C₅H₆ClN₃
MW: 143.58 g/mol
MDL Number: MFCD16251142
CAS Number: 145934-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR