N-(2-Hydrazino-2-oxoethyl)benzamide

Supplier: Matrix Scientific

077575-500MG 077575-1G 077575-5G
102666-948EA 258.46 USD
102666-948 102666-950 102666-952
N-(2-Hydrazino-2-oxoethyl)benzamide
N-(2-Hydrazino-2-oxoethyl)benzamide

Hazard: Irritant

Formula: C₉H₁₁N₃O₂
MDL Number: MFCD00462266
CAS Number: 2443-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR