tert-Butyl-4-(2-methoxy-2-oxoethyl)piperazine-1-carboxylate

Supplier: Matrix Scientific

077573-500MG 077573-1G 077573-5G
102666-936EA 207.69 USD
102666-936 102666-938 102666-940
tert-Butyl-4-(2-methoxy-2-oxoethyl)piperazine-1-carboxylate
tert-Butyl-4-(2-methoxy-2-oxoethyl)piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₄
MDL Number: MFCD02684439
CAS Number: 731810-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR