tert-Butyl-4-(2-amino-2-oxoethyl)piperazine-1-carboxylate
Supplier: Matrix Scientific
077571-500MG
077571-1G
077571-5G
102666-926EA
258.46
USD
102666-926
102666-928
102666-930
tert-Butyl-4-(2-amino-2-oxoethyl)piperazine-1-carboxylate
tert-Butyl-4-(2-amino-2-oxoethyl)piperazine-1-carboxylate
Hazard: Irritant
Formula:
C₁₁H₂₁N₃O₃ MW: 243.31 g/mol |
MDL Number:
MFCD08062583 CAS Number: 77278-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...