4-(3-Bromophenyl)-4H-1,2,4-triazole

Supplier: Matrix Scientific

077553-500MG 077553-1G 077553-5G
102666-842EA 387.69 USD
102666-842 102666-844 102666-846
4-(3-Bromophenyl)-4H-1,2,4-triazole
4-(3-Bromophenyl)-4H-1,2,4-triazole

Hazard: Irritant

Formula: C₈H₆BrN₃
MDL Number: MFCD09971233
CAS Number: 375858-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR