N'-Acetyl-2-chloroacetohydrazide

Supplier: Matrix Scientific

077551-500MG 077551-1G
102666-834EA 258.46 USD
102666-834 102666-836
N'-Acetyl-2-chloroacetohydrazide
N'-Acetyl-2-chloroacetohydrazide

Hazard: Irritant

Formula: C₄H₇ClN₂O₂
MDL Number: MFCD04035423
CAS Number: 4002-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR