5-Isopropyl-1,3,4-thiadiazol-2-ol
Supplier: Matrix Scientific
077549-500MG
077549-1G
077549-5G
102666-822EA
516.92
USD
102666-822
102666-824
102666-826
5-Isopropyl-1,3,4-thiadiazol-2-ol
5-Isopropyl-1,3,4-thiadiazol-2-ol
Hazard: Irritant
Formula:
C₅H₈N₂OS |
MDL Number:
MFCD19982758 CAS Number: 84352-67-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...