3-Amino-2-methylpropan-1-ol

Supplier: Matrix Scientific

077544-500MG 077544-1G 077544-5G
102666-796EA 292.31 USD
102666-796 102666-798 102666-800
3-Amino-2-methylpropan-1-ol
3-Amino-2-methylpropan-1-ol

Hazard: Irritant

Formula: C₄H₁₁NO
MDL Number: MFCD03412698
CAS Number: 15518-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR