4,5-Dimethyl-1,3-oxazol-2-amine hydrochloride
Supplier: Matrix Scientific
077543-500MG
077543-1G
077543-5G
102666-790EA
646.15
USD
102666-790
102666-792
102666-794
4,5-Dimethyl-1,3-oxazol-2-amine hydrochloride
4,5-Dimethyl-1,3-oxazol-2-amine hydrochloride
Hazard: Irritant
Formula:
C₅H₉ClN₂O |
MDL Number:
MFCD20441426 CAS Number: 45529-92-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...