3-(Benzylamino)-2-methylpropan-1-ol

Supplier: Matrix Scientific

077538-500MG 077538-1G 077538-5G
102666-776EA 387.69 USD
102666-776 102666-778 102666-780
3-(Benzylamino)-2-methylpropan-1-ol
3-(Benzylamino)-2-methylpropan-1-ol

Hazard: Irritant

Formula: C₁₁H₁₇NO
MDL Number: MFCD08691880
CAS Number: 858834-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR